Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc cyanide, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
Packaging:
Glass Bottle
Plastic bottle
557-21-1
C2N2Zn
117.43
MFCD00011292
557-21-1
C2N2Zn
117.43
MFCD00011292
C-X bond formation reagent
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula Zn(CN)2
Beilstein 02,III,86
Merck Index 15,10334
Solubility Information Solubility in water: insoluble
Formula Weight 117.43
Physical Form Powder
Percent Purity 98%
Grade Pure
Packaging Glass Bottle
Solubility Solubility in water: insoluble.
Color White
Melting Point 800.0°C
Quantity 5g
Chemical Name or Material Zinc cyanide, 98%
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Contact with acids liberates very toxic gas.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-162-9