Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc finger protein 395 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Zinc finger protein 395 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Zinc finger protein 395 Polyclonal specifically detects Zinc finger protein 395 in Human samples. It is validated for Western Blot.Specifications
Zinc finger protein 395 | |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
PBS buffer, 2% sucrose | |
55893 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
dJ874C20.2, FLJ23407, zinc finger protein 310 pseudogene, zinc finger protein 323, ZNF20-Lp, ZNF310P | |
The immunogen is a synthetic peptide directed towards the middle region of human Zinc finger protein 395 (NP_061130). Peptide sequence HLIVTSPPRAQSGARKARGEAKKCRKVYGIEHRDQWCTACRWKKACQRFL | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title