Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc finger protein 754 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987212
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7212 Supplier Bioss Supplier No. BS6347R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Zinc finger proteins have regions (zinc finger domains) consisting of cysteines and histidines or cysteines alone which can form a tetrahedral complex around a Zinc ion. Zinc finger represent a class of DNA-binding proteins, act as transcriptional regulators of other genes. These multifunctional transcription factors exhibits control on a large number of cellular genes by binding to sites overlapping the transcription start site and plays an important role in development and differentiation. Hromas et al. in an effort to identify activators of the genetic cascade in hemopoietic differentiation probed a human myeloid cDNA library. ZNF42 may be a regulator of transcriptional events during hemopoietic development.

Specifications

Antigen Zinc finger protein 754
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZFP42
Gene Accession No. Q96MM3
Gene Alias hREX-1; mREX-1; Reduced expression protein 1; REX1; Rex-1; REX1 transcription factor; ZFP42; Zfp-42; ZFP42 zinc finger protein; Zfp971; Zinc finger protein 42; Zinc finger protein 42 homolog; Zinc finger protein 754; zinc finger protein 971; ZNF754
Gene Symbols ZFP42
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Rex1/Zinc finger protein 754.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132625
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.