Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc finger protein 774 Antibody (PCRP-ZNF774-3F7), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320746MFV500
Description
Zinc finger protein 774 Monoclonal antibody specifically detects Zinc finger protein 774 in Human samples. It is validated for Flow CytometrySpecifications
Zinc finger protein 774 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
342132 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
PCRP-ZNF774-3F7 | |
Flow Cytometry | |
MGC75360, zinc finger protein 774 | |
Recombinant full-length human Zinc finger protein 774 protein | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction