Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc finger protein 774 Antibody (PCRP-ZNF774-3F7), Novus Biologicals™
SDP

Catalog No. NB235397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB235397 20 μg
NB235394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB235397 Supplier Novus Biologicals Supplier No. NBP32047220UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zinc finger protein 774 Monoclonal antibody specifically detects Zinc finger protein 774 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Zinc finger protein 774
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone PCRP-ZNF774-3F7
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-2 μg/million cells, ELISA
Formulation 10mM PBS with 0.05% BSA
Gene Alias MGC75360, zinc finger protein 774
Host Species Mouse
Immunogen Recombinant full-length human Zinc finger protein 774 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342132
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.