Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc finger protein 790 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2838120.1ML
Description
Zinc finger protein 790 Polyclonal specifically detects Zinc finger protein 790 in Human samples. It is validated for Western Blot.Specifications
| Zinc finger protein 790 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 388536 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| FLJ20350, MGC62100, zinc finger protein 790 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human Zinc finger protein 790 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction