Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Flake, -325 mesh, 97% (Metals basis), Thermo Scientific™

Catalog No. p-4782235
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray flake
Quantity 500 g
Mesh Size -325 Mesh
Odor Odorless
Form Flake
Assay Percent Range 97% (Metals basis)
Chemical Name or Material Zinc
Hazard Category H250-H260-H500
Hazard Statement If swallowed seek medical advice immediately and show this container or label. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-175-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.