Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc fluoride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
Packaging:
Plastic bottle
7783-49-5
F2Zn
103.38
MFCD00011298
BHHYHSUAOQUXJK-UHFFFAOYSA-L
zinc ii fluoride,zinc 2+ ion difluoride,acmc-1bhy1,ksc379c0l,zinc fluoride 50g,zinc, ion zn2+ difluoride
10129912
zinc;difluoride
[F-].[F-].[Zn+2]
7783-49-5
F2Zn
103.38
MFCD00011298
BHHYHSUAOQUXJK-UHFFFAOYSA-L
zinc ii fluoride,zinc 2+ ion difluoride,acmc-1bhy1,ksc379c0l,zinc fluoride 50g,zinc, ion zn2+ difluoride
10129912
zinc;difluoride
[F-].[F-].[Zn+2]
Assay Percent Range 98%
Linear Formula ZnF2
Fieser 06,514
Merck Index 15, 10335
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in hcl,hno3 and nh4oh,insoluble in alcohol
Formula Weight 103.38
Physical Form Powder or Crystals
Percent Purity 98%
Packaging Plastic bottle
Solubility Solubility in water: slightly soluble. Other solubilities: freely soluble in hcl, hno3 and nh4oh, insoluble in alcohol
Zinc (Zn) 61.7 to 64.8 %
Boiling Point 1500.0°C
Color Brown to White
Melting Point 872.0°C to 910.0°C
Quantity 25g
Chemical Name or Material Zinc fluoride, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 232-001-9

RTECSNumber : ZH3500000

TSCA : TSCA