Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Fluoride, Hydrate 99% (ca 4 Mole H2O), Thermo Scientific™

Catalog No. AC299780500
Click to view available options
Quantity:
50 g
Packaging:
Plastic Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13986-18-0
Molecular Formula F2H8O4Zn
Molecular Weight (g/mol) 175.437
InChI Key OEUUYPRXVRBIFO-UHFFFAOYSA-L
Synonym zinc fluoride tetrahydrate, zinc fluoride, tetrahydrate, unii-sq7815wxfb, zinc difluoride tetrahydrate, sq7815wxfb, zinc fluoride tetrahydrate mi, acmc-1c1qo, ksc497c6d, zinc, ion zn2+ tetrahydrate difluoride, zinc fluoride, tetrahydrate,
PubChem CID 44717730
IUPAC Name zinc;difluoride;tetrahydrate
SMILES O.O.O.O.[F-].[F-].[Zn+2]

Specifications

Quantity 50 g
Linear Formula ZnF2.4H2O
Formula Weight 175.44
Packaging Plastic Bottle
Chemical Name or Material Zinc fluoride, hydrate, 99% (ca 4 mole H2O)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.