Learn More
Zinc fluoride, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Description
It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1500°C |
Melting Point | 872°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | ZnF2 |
Merck Index | 14,10136 |
Solubility Information | Sparingly soluble in hydrochloric acid,nitric acid,and ammonium hydroxide. |
Formula Weight | 103.37 |
Percent Purity | 99.995% |
Show More |
Safety and Handling
Hazard Category | H315-H318-H335 |
Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
EINECSNumber | 232-001-9 |
RTECSNumber | ZH3500000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.