Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc fluoride, Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 1298606

Quantity 5g

 View more versions of this product

Catalog No. AA1298606


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7783-49-5
103.37
14,10136
Puratronic™
ZnF2
MFCD00011298
Zinc fluoride

Specifications

7783-49-5
ZnF2
MFCD00011298
4.95g/mL
White
5g
99.995%, (metals basis)
Powder
103.37
14,10136
1500°C
872°C
Zinc fluoride
Puratronic™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : ZH3500000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit