Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc fluoride tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog No. p-7049356
Click to view available options
Quantity:
500 g

Zinc fluoride tetrahydrate is used as a mild fluorinating agent in the replacement of chlorine in halogenated hydrocarbons, in fluorophosphate and fluoride glass, and in the manufacture of fluoride glass optical fibers and optical transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13986-18-0
Molecular Formula F2H8O4Zn
Molecular Weight (g/mol) 175.437
MDL Number MFCD00149892
InChI Key OEUUYPRXVRBIFO-UHFFFAOYSA-L
Synonym zinc fluoride tetrahydrate, zinc fluoride, tetrahydrate, unii-sq7815wxfb, zinc difluoride tetrahydrate, sq7815wxfb, zinc fluoride tetrahydrate mi, acmc-1c1qo, ksc497c6d, zinc, ion zn2+ tetrahydrate difluoride, zinc fluoride, tetrahydrate,
PubChem CID 44717730
IUPAC Name zinc;difluoride;tetrahydrate
SMILES O.O.O.O.[F-].[F-].[Zn+2]

Specifications

Physical Form Crystalline
Quantity 500 g
UN Number UN3288
Merck Index 14,10136
Solubility Information Slightly soluble in water
Formula Weight 175.43 (103.37 Anhydrous)
Percent Purity 98%
Odor Odorless
Chemical Name or Material Zinc fluoride tetrahydrate
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 232-001-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.