Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc formate, 98%

Catalog No. p-7027452
Click to view available options
Quantity:
100 g
500 g

CAS: 557-41-5 | C2H2O4Zn | 155.414 g/mol

Zinc formate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc formate is used as a pharmaceutical intermediate.

Solubility
Soluble in water (52 g/L at 20°C).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-41-5
Molecular Formula C2H2O4Zn
Molecular Weight (g/mol) 155.414
MDL Number MFCD00050836
InChI Key XOBMCBQSUCOAOC-UHFFFAOYSA-L
Synonym zinc formate, zinc diformate, formic acid, zinc salt, caswell no. 915, unii-zz67g2ct75, formic acid, zinc salt 2:1, epa pesticide chemical code 087802, 64-18-6 parent, bis-formyloxy zinc, zinc 2+ diformate
PubChem CID 11195
IUPAC Name zinc;diformate
SMILES C(=O)[O-].C(=O)[O-].[Zn+2]

Specifications

Quantity 100 g
UN Number UN3077
Solubility Information Soluble in water (52g/L at 20°C).
Formula Weight 155.4
Percent Purity 98%
Chemical Name or Material Zinc formate
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 209-176-5
RTECSNumber LR0725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.