Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc gluconate hydrate, 97%

Catalog No. aab2302236
Click to view available options
Quantity:
100 g
500 g

CAS: 4468-02-4 | C4H7NO | 85.11 g/mol

Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

Solubility
Soluble in water 100g/ l.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4468-02-4
MDL Number MFCD00868110

Specifications

pH 6.5
Odor Odorless
Molecular Formula C4H7NO
Quantity 500 g
Merck Index 14,4456
Synonym propionaldehyde cyanohydrin, 2-hydroxybutyronitrile, butyronitrile, 2-hydroxy, butanenitrile, 2-hydroxy, alpha-hydroxybutyronitrile, propanal cyanhydrin, 1-cyanopropan-1-ol, 2-oxidanylbutanenitrile, 2-hydroxy-butyronitrile, butanenitrile, 2-hydroxy-9ci
Solubility Information Soluble in water 100g/ l.
InChI Key NHSSTOSZJANVEV-UHFFFAOYNA-N
SMILES CCC(C#N)O
IUPAC Name 2-hydroxybutanenitrile
Molecular Weight (g/mol) 85.11
PubChem CID 20553
Formula Weight 455.69 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Zinc gluconate hydrate
Show More Show Less
EINECSNumber 224-736-9
RTECSNumber ZH3750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.