Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Gluconate, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18607852 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-607-852

Supplier: Spectrum Chemical Manufacturing Corporation Z107712KG

Only null left
Add to Cart
Add to Cart

Z1077, 4468-02-4, C12H22O14Zn

Spectrum™ Chemical Zinc Gluconate, USP is used for treatment and prevention of zinc deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4468-02-4
Molecular Formula C12H22O14Zn
Molecular Weight (g/mol) 455.67
MDL Number MFCD00868110
InChI Key WHMDKBIGKVEYHS-IYEMJOQQSA-L
IUPAC Name zinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 1
pH 5.5 to 7.5
Packaging Poly Pail
Quantity 12 kg
Percent Purity 97 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.