Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc ICP Standard, 10,000 ppm Zn in 5% HNO3, Ricca Chemical
SDP

Catalog No. PZN10KN100
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Chemical Identifiers

CAS 7440-66-6, 7697-37-2, 7732-18-5
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Zn
CAS Min % 87.0, 10.42, 0.87
CAS Max % 90.0, 10.85, 0.9
Equivalence 1 mL = 10 mg Zn
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Zinc ICP Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.