Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Zinc iodide, 99.999%, (trace metal basis), extra pure
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                5 g
                            
                        
                            
                                25 g
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Glass bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
        
            
            Specifications
Specifications
| Melting Point | 446°C | 
| Quantity | 25 g | 
| Assay Percent Range | 99.999% | 
| Linear Formula | ZnI2 | 
| Fieser | 01,1294; 05,765; 10,462; 11,604; 12,576; 13,88 | 
| Merck Index | 15, 10339 | 
| Solubility Information | Solubility in water: 4500g/L (20°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether | 
| Formula Weight | 319.19 | 
| Percent Purity | 100.00% | 
| Grade | Trace Metal Basis | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Danger | 
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage.  | 
                                
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi  | 
                                
| EINECSNumber | 233-396-0 | 
| TSCA | TSCA | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction