Learn More
Zinc iodide, ultra dry, 99.995% (metals basis)
Description
Zinc iodide is used as an x-ray opaque penetrant in industrial radiography to enhance the contrast. It is used as electrode compartments and as a stain in electron microscopy in association with osmium tetroxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 625°C (decomposition) |
| Melting Point | 446°C |
| Physical Form | Granules |
| Quantity | 100 g |
| Assay Percent Range | 99.9%, metal basis |
| Solubility Information | Soluble in water. |
| Formula Weight | 319.18 |
| Sensitivity | Hygroscopic |
| Density | 4.74 |
| Chemical Name or Material | Zinc Iodide, Ampouled under argon |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | DOT Class: 8; Packaging Group: II |
| EINECSNumber | 233-396-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.