Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc molybdenum oxide, 98%

Catalog No. p-7080514
Click to view available options
Quantity:
5 g
25 g

CAS: 13767-32-3 | ZnMoO4

Used as a corrosion inhibitor in paints, adhesives, primer on metal surfaces, such as steel and aluminum. It may be applied by brush or spray, and is suitable as an after-blast or after-pickling, primer for plating, as an undercoat for alkyd enamels. Also used for dielectric and insulation applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13767-32-3
Molecular Formula ZnMoO4
MDL Number MFCD00049622
Synonym Zinc molybdate

Specifications

Melting Point >700°C
Physical Form Powder
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 225.32
Percent Purity 98%
Chemical Name or Material Zinc molybdenum oxide
EINECSNumber 237-377-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.