Learn More
Zinc molybdenum oxide, 98%, Thermo Scientific Chemicals
Description
Used as a corrosion inhibitor in paints, adhesives, primer on metal surfaces, such as steel and aluminum. It may be applied by brush or spray, and is suitable as an after-blast or after-pickling, primer for plating, as an undercoat for alkyd enamels. Also used for dielectric and insulation applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >700°C |
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 225.32 |
Percent Purity | 98% |
Chemical Name or Material | Zinc molybdenum oxide |
Safety and Handling
EINECSNumber | 237-377-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.