Zinc Oxide, 99.999% (Metal Basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1113730

Quantity 250g

 View more versions of this product

Catalog No. AA1113730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-13-2
    Physical Form Powder
    Assay Percent Range 99.999%, metal basis
    Molecular Formula ZnO
    Formula Weight 81.37
    MDL Number MFCD00011300
    Chemical Name or Material Zinc Oxide

    Specifications

    Formula Weight 81.37
    Quantity 250g
    DOT Information DOT Class: 9
    EINECS Number 215-222-5
    Physical Form Powder
    Assay Percent Range 99.999%, metal basis
    Molecular Formula ZnO
    MDL Number MFCD00011300
    CAS 1314-13-2
    Health Hazard 1 Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
    Health Hazard 2 This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets
    Chemical Name or Material Zinc Oxide

    This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets

    Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit