Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Oxide, ACS,Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1088491000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
25 kg
Packaging:
Fibre carton
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1088491000 1 kg Plastic bottle
M1088490500 500 g Plastic bottle
M1088499025 25 kg Fibre carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1088491000 Supplier MilliporeSigma Supplier No. 1088491000
Add to Cart
Add to Cart
For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Physical Form Solid
Quantity 1 kg
UN Number UN 3077
Formula Weight 81.37. (molar mass)
Grade ACS/Reagent Ph. Eur.
Odor Odorless
Packaging Plastic bottle
Appearance White solid
Chemical Name or Material Zinc oxide
Hazard Category H410: Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-222-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.