Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Oxide, NanoGard™, Thermo Scientific™

Catalog No. AA4489822
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4489822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4489822 Supplier Thermo Scientific Chemicals Supplier No. 4489822

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Physical Form Powder
Quantity 100 g
Formula Weight 81.37
Chemical Name or Material Zinc Oxide
Hazard Category Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
Hazard Statement This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.