Learn More
Zinc oxide, NanoShield™ ZN-2000, 50% in H2O, colloidal dispersion with nonionic dispersant, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Zinc oxide water borne dispersion suitable for incorporation in water compatible coatings and paints
Specifications
Specifications
| Physical Form | Dry Powder |
| Quantity | 100 g |
| UN Number | UN3082 |
| Formula Weight | 81.37 |
| Concentration or Composition (by Analyte or Components) | 50% in H2O |
| Odor | Odorless |
| Chemical Name or Material | Zinc oxide, NanoShield™ ZN-2000, Colloidal dispersion with nonionic dispersant |
Safety and Handling
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
| TSCA | TSCA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.