Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc oxide, NanoTek™ C2, 99%, organosilane coated, hydrophobic, polar, Thermo Scientific™

Catalog No. AA4490136
Click to view available options
Quantity:
500g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • NanoTek™ zinc oxide C2 has an oleophilic, polar coating to aid dispersing in organic systems
TRUSTED_SUSTAINABILITY

Specifications

CAS 1314-13-2
Quantity 500g
Molecular Formula OZn
MDL Number MFCD00011300
UN Number UN3077
Merck Index 14,10147
Synonym zinc oxide, zinc white, amalox, permanent white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILES O=[Zn]
IUPAC Name oxozinc
Molecular Weight (g/mol) 81.379
PubChem CID 14806
ChEBI CHEBI:36560
Formula Weight 81.37
Percent Purity 99%
Odor Odorless
Chemical Name or Material Zinc oxide, NanoTek™ C2, Organosilane coated, Hydrophobic, Polar
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECSNumber ZH4810000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.