Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Oxide, puriss. p.a., ACS Reagent, ≥99% (KT), Solstice

Catalog No. 60047788 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-788 100 g Poly Bottle
60-047-791 500 g Poly Bottle
60-047-790 250 g Poly Bottle
60-047-789 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-047-788 Supplier Solstice Supplier No. 96479100G

Puriss. p.a., ACS Reagent,≥99.0% (KT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point ∼1975°C
Color White
pH ∼7 (at 20°C)
Quantity 100 g
Linear Formula ZnO
UN Number UN3077
Formula Weight 81.39g/mol
Percent Purity ≥99% (KT)
Packaging Poly Bottle
Appearance Powder
Density 5.6g/cm3 at 20°C
Chemical Name or Material Zinc oxide
Show More Show Less
Hazard Statement P273-P501
DOTInformation 9/PGIII
RTECSNumber 215-222-5
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.