Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc perchlorate hexahydrate, 99.997% (metals basis)

Catalog No. p-7059277
Click to view available options
Quantity:
10 g
50 g

CAS: 10025-64-6 | Cl2H12O14Zn | 372.362 g/mol

Used as intermediates. A synthetic reagent for proteomics research. Zinc(II) perchlorate hexahydrate is a highly efficient catalyst for the opening of epoxides by amines affording 2-amino alcohols in high yields under solvent-free conditions and with excellent chemo-, regio-, and stereoselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-64-6
Molecular Formula Cl2H12O14Zn
Molecular Weight (g/mol) 372.362
MDL Number MFCD00150370
InChI Key PADPILQDYPIHQQ-UHFFFAOYSA-L
Synonym zinc perchlorate hexahydrate, perchloric acid, zinc salt, hexahydrate, 2clo4.zn.6h2o, zinc diperchlorate hexahydrate, zinc perchlorate 6h2o, zincperchloratehexahydrate, zinc 2+ hexahydrate diperchlorate, zinc perchlorate hexahydrate, reagent grade, zinc, ion zn2+ hexahydrate diperchlorate ion, zinc perchlorate hexahydrate trace metals basis 10g
PubChem CID 202253
IUPAC Name zinc;diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Zn+2]

Specifications

Melting Point 106°C
Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula Zn(ClO4)2·6H2O
UN Number UN1481
Merck Index 14,10148
Solubility Information Soluble in ethyl alcohol and water.
Formula Weight 372.37 (264.28 Anhydrous)
Percent Purity 99.997%
Odor Odorless
Refractive Index 1.5
Density 2.25 g/mL
Assay (metals basis)
Chemical Name or Material Zinc perchlorate hexahydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 237-122-0
RTECSNumber SD0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.