Alfa Aesar™ Zinc perchlorate hexahydrate, 99.997% (metals basis)

Manufacturer: Alfa Aesar™ 4431509

Quantity 10g

 View more versions of this product

Catalog No. AA4431509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-64-6
    Molecular Formula Cl2O8Zn·6H2O
    Linear Formula Zn(ClO4)2·6H2O
    Formula Weight 372.37 (264.28 Anhydrous)
    MDL Number MFCD00150370
    UN Number UN1481
    Merck Index 14,10148
    Chemical Name or Material Zinc perchlorate hexahydrate

    Specifications

    Formula Weight 372.37 (264.28 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula Cl2O8Zn·6H2O
    MDL Number MFCD00150370
    Health Hazard 1 Danger
    Refractive Index 1.50
    Melting Point 106°C
    Odor Odorless
    Physical Form Crystalline
    CAS 10025-64-6
    UN Number UN1481
    Color White
    Quantity 10g
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
    RTECS Number SD0525000
    TSCA TSCA
    Linear Formula Zn(ClO4)2·6H2O
    Merck Index 14,10148
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 2.25g/mL
    Chemical Name or Material Zinc perchlorate hexahydrate
    Assay 99.997%, (metals basis)

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit