Alfa Aesar™ Zinc perchlorate hexahydrate, Reagent Grade

Manufacturer: Alfa Aesar™ 1161422

Quantity 100g

 View more versions of this product

Catalog No. AA1161422

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 10025-64-6
    Linear Formula Zn(ClO4)2·6H2O
    Formula Weight 372.37 (264.28 Anhydrous)
    MDL Number MFCD00150370
    UN Number UN1481
    Merck Index 14,10148
    Chemical Name or Material Zinc perchlorate hexahydrate
    Grade Reagent Grade

    Specifications

    Formula Weight 372.37 (264.28 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Physical Form Crystalline
    Sensitivity Hygroscopic
    MDL Number MFCD00150370
    CAS 10025-64-6
    UN Number UN1481
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 2.25g/mL
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
    TSCA TSCA
    Refractive Index 1.50
    Linear Formula Zn(ClO4)2·6H2O
    Merck Index 14,10148
    Quantity 100g
    Solubility Information Soluble in water, alcohol
    Health Hazard 1 Danger
    RTECS Number SD0525000
    Color White
    Melting Point 106°C
    Chemical Name or Material Zinc perchlorate hexahydrate
    Grade Reagent Grade

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit