Learn More
Zinc phosphide, 99.999% (metals basis)
Description
Zinc phosphide has useful applications in photovoltaic cells. It is also employed as a laboratory chemical. It plays an important role in medicine and pharmaceuticals. It acts as a fumigant in grain storage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1100°C |
Melting Point | 420°C |
Physical Form | Pieces |
Quantity | 0.5 g |
UN Number | UN1714 |
Merck Index | 14,10152 |
Solubility Information | Insoluble in water and ethanol. Soluble in benzene. |
Formula Weight | 258.1 |
Percent Purity | 99.999% |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H260-H300 |
Hazard Statement | GHS H Statement H260-H300 In contact with water releases flammable gases which may ignite spontaneously. Fatal if swallowed. |
Precautionary Statement | P223-P231+P232-P264b-P270-P280-P301+P310-P330-P335+P334-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ZINC PHOSPHIDE |
EINECSNumber | 215-244-5 |
RTECSNumber | ZH4900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.