Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc shot, 1-6mm (0.04-0.24in), Puratronic™, 99.9999% (metals basis)

Catalog No. p-3257062
Click to view available options
Quantity:
50 g
250 g
1 kg

CAS: 7440-66-6 | Zn | 65.38 g/mol

Polytetrafluoroethylene, 6-10 micron is used in specially designed coating applications on coil for bakeware.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray shot
Density 7.14g/cm3
Quantity 50 g
Odor Odorless
Form Shot
Assay Percent Range 99.9999% (Metals basis)
Formula Weight 65.3
Chemical Name or Material Zinc, 1-6mm (0.04-0.24 in.)
EINECSNumber 231-175-3
RTECSNumber ZG8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.