Zinc shot, 1-6mm (0.04-0.24in), Puratronic, 99.9999% (metals basis), Alfa Aesar

Manufacturer: Alfa Aesar 1076018

Quantity 50g

 View more versions of this product

Catalog No. AA1076018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-66-6
    Physical Form Shot, 1 To 6mm (0.04 To 0.24 in.)
    Assay Percent Range 99.9999% (Metals basis)
    Molecular Formula Zn
    Formula Weight 65.3
    MDL Number MFCD00011291
    Chemical Name or Material Zinc, 1-6mm (0.04-0.24 in.)

    Specifications

    Physical Form Shot, 1 To 6mm (0.04 To 0.24 in.)
    Formula Weight 65.3
    Quantity 50g
    Color Gray
    Density 7.14g/cm3
    EINECS Number 231-175-3
    Assay Percent Range 99.9999% (Metals basis)
    Molecular Formula Zn
    MDL Number MFCD00011291
    CAS 7440-66-6
    Odor Odorless
    Appearance Gray shot
    Chemical Name or Material Zinc, 1-6mm (0.04-0.24 in.)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit