Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Standard, 1mL = 1mg Zn, 1000ppm Zn, Ricca Chemical
SDP

Catalog No. 95004 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Poly natural
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7440-66-6, 7697-37-2, 7732-18-5
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Packaging Poly natural
Concentration or Composition Notes 1,000ppm Zn
Equivalence 1 mL = 1 mg Zn
Physical Form Liquid
Color Colorless
Quantity 120 mL
Chemical Name or Material Zinc Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.