Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Std, 1.5 mg/L, Reagents

Catalog No. 01333359
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-359 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-359 Supplier Reagents Holdings Llc Supplier No. CZ0015001B
Only null left
Add to Cart
Add to Cart

Zinc Std, 1.5 mg/L, Reagent, use for Zinc analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7646-85-7 , 7732-18-5
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
IUPAC Name zinc(2+) dichloride
SMILES [Cl-].[Cl-].[Zn++]

Specifications

Packaging Opaque HDPE
Specific Gravity 1
Physical Form Liquid
Color Colorless
Boiling Point 102°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Chemical Name or Material Zinc Std, 1.5 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.