Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Std, 500 mg/L, Reagents

Catalog No. 01333357
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-357 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-357 Supplier Reagents Holdings Llc Supplier No. CZ0050001B

Due to product restrictions, please to purchase or view availability for this product.

Zinc Std, 500 mg/L, Reagent, use for Zinc analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7646-85-7 , 7732-18-5
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
IUPAC Name zinc(2+) dichloride
SMILES [Cl-].[Cl-].[Zn++]

Specifications

Packaging Opaque HDPE
Specific Gravity 1
Physical Form Liquid
Color Colorless
Boiling Point 102°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Quantity 1 L
Percent Purity 0.05%
Grade Reagent
Chemical Name or Material Zinc Std, 500 mg/L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.