Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc stearate, ZnO 12.5-14%

Catalog No. AA33238A7
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg

CAS: 557-05-1 | C36H70O4Zn | 632.332 g/mol

Zinc stearate is used as a release agent for the manufacturing of rubber, polyurethane and polyester processing. It is a lubricant and thickening agent in cosmetic industry. It plays a vital role as an activator for rubber vulcanization. It acts as a phase transfer catalyst in the saponification of fats due to its lipophilic character. It is an active component in fanning powder, which is utilized to decrease the friction between cards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-05-1
Molecular Formula C36H70O4Zn
Molecular Weight (g/mol) 632.332
MDL Number MFCD00013031
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
Synonym zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat
PubChem CID 11178
IUPAC Name zinc;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2]

Specifications

Melting Point 130°C
Color White
Quantity 5 kg
Merck Index 14,10158
Solubility Information Insoluble in water,alcohol and ether; Soluble in benzeneSoluble in acids,aromatic and chlorinated hydrocarbons; Insoluble in water,alcohol and ether.
Formula Weight 632.48
Physical Form Powder
Chemical Name or Material Zinc stearate
Hazard Category H500
EINECSNumber 209-151-9
RTECSNumber WI4390000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.