Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc sulfate heptahydrate, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
7446-20-0
O4SZn·7H2O
287.53
MFCD00149894
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc sulfate heptahydrate,zinc sulfate jan,zinc sulfate 1:1 heptahydrate,unii-n57ji2k7wp,zinc vitriol heptahydrate,zinc sulfate heptahydrate 1:1:7,white vitriol heptahydrate,ccris 5563,zinc sulfate znso4 heptahydrate,znso4.7h2o
62640
CHEBI:32312
zinc;sulfate;heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
7446-20-0
O4SZn·7H2O
287.53
MFCD00149894
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc sulfate heptahydrate,zinc sulfate jan,zinc sulfate 1:1 heptahydrate,unii-n57ji2k7wp,zinc vitriol heptahydrate,zinc sulfate heptahydrate 1:1:7,white vitriol heptahydrate,ccris 5563,zinc sulfate znso4 heptahydrate,znso4.7h2o
62640
CHEBI:32312
zinc;sulfate;heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Zn+2]
Assay Percent Range 99.4% min. (Complexometry)
Linear Formula ZnSO4·7H2O
Merck Index 15, 10359
Arsenic (As) 1ppm max.
Solubility Information Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol,insoluble in alcohol
Formula Weight 287.53
Physical Form Crystals or Crystalline Powder
Percent Purity 99.5%
Grade Analysis
Chloride 5ppm max
Chlorine (Cl) 5ppm max.
Iron (Fe) 10ppm max.
Lead (Pb) 10ppm max.
Manganese (Mn) 3ppm max.
Nitrogen Compounds 0.002% max.
Packaging Plastic bottle
Potassium (K) 10ppm max.
Sodium (Na) 10ppm max.
Solubility Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol, insoluble in alcohol
Color White
Melting Point 100.0°C
pH 4.4 to 6 (5% soln. at 20°C)
Quantity 250g
Chemical Name or Material Zinc sulfate heptahydrate
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger