Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate Heptahydrate, ACS Reagent, Reagents

Catalog No. p-200020971
Click to view available options
Quantity:
100 g
500 g
25 kg

Catalog No. 01-337-623

Supplier: Reagents Holdings Llc C1354000100C2

Only null left
Add to Cart
Add to Cart

Zinc Sulfate Heptahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym Zinc Sulphate Heptahydrate
IUPAC Name zinc(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Boiling Point >500°C
Melting Point 100°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.97
Chemical Name or Material Zinc Sulfate Heptahydrate, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.