Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc sulfate hydrate, Puratronic, 99.998% (metals basis)

Catalog No. AA1113818
Click to view available options
Quantity:
10 g
50 g

CAS: 16788-42-4 | H2O5SZn | 179.45 g/mol

Zinc sulfate hydrate is used in the production of rayon, as a feed supplement, and as a fertilizer ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16788-42-4
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.45
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
SMILES O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 100°C
Color White
Physical Form Crystalline Powder
Quantity 50 g
Linear Formula ZnSO4·xH2O
UN Number UN3077
Merck Index 14,10159
Solubility Information Soluble in water.
IUPAC Name zinc(2+) hydrate sulfate
Formula Weight 161.43 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc sulfate hydrate
Show More Show Less
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-793-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.