Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate, 1.180 Specific Gravity, for Concentration of Intestinal Parasites, Eggs, and Cysts, Ricca Chemical
SDP

Catalog No. 96501 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle

Catalog No. 9650-1

Supplier: Ricca Chemical Company 96501

Only null left
Add to Cart
Add to Cart

For Concentration of Intestinal Parasites, Eggs, and Cysts

Chemical Identifiers

CAS 7446-20-0, 7732-18-5
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

CAS Min % 74.0, 24.31
CAS Max % 77.0, 25.3
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous solution
pH ∼4.5
Quantity 4 L
Chemical Name or Material Zinc Sulfate Solution, 1.180 Specific Gravity
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.