Learn More
Zinc sulfide, 99.9% (metals basis)
Description
Zinc sulfide is used in luminescent materials, infra red optical materials, semiconductors, as a pigment, and as a window for visible optics and infrared optics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1700°C |
| Physical Form | Pieces |
| Quantity | 250 g |
| Assay Percent Range | 99.9%, metal basis |
| Solubility Information | Soluble in acids. Insoluble in water. |
| IUPAC Name | sulfanylidenezinc |
| Formula Weight | 97.43 |
| Density | 4.1 |
| Chemical Name or Material | Zinc Sulfide |
Safety and Handling
| EINECSNumber | 215-251-3 |
| RTECSNumber | ZH5400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.