Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc tetrafluoroborate hydrate, Zn 18% min

Catalog No. p-4782243
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 27860-83-9 | B2F8Zn | 238.99 g/mol

Zinc tetrafluoroborate hydrate is used as a precursor for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties. Further, it serves as a mild catalyst for epoxide ring opening with amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27860-83-9
Molecular Formula B2F8Zn
Molecular Weight (g/mol) 238.99
MDL Number MFCD00150371
InChI Key BUMIVIYHDVNOQE-UHFFFAOYSA-N
Synonym zinc tetrafluoroborate hydrate, 2bf4.zn.h2o, zinc tetrafluoroborate-water 1/2/1, zinc 2+ hydrate ditetrafluoroborate, zinc 2+ ion hydrate ditetrafluoroborate, borate 1-,tetrafluoro-, zinc 2:1 , hexahydrate 9ci
PubChem CID 16212112
SMILES [Zn++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Soluble in water and alcohol.
IUPAC Name zinc;ditetrafluoroborate;hydrate
Formula Weight 238.98 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Zinc Tetrafluoroborate
Hazard Category H314-H335
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 237-534-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.