Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc transporter 9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Zinc transporter 9 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Zinc transporter 9 Polyclonal specifically detects Zinc transporter 9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Zinc transporter 9 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10463 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:HRCSWSSLCRLRLRCRAAACNPSDRQEWQNLVTFGSFSNVVPCSHPYIGTLSQVKLYSTNVQKEGQGSQTLRVEKVPSFETAEGIGAELKAPLKQEPLQVRVKAVLKKREYGSKYTQNNFITGV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
C4orf1GRIP1-dependent nuclear receptor coactivator, chromosome 4 open reading frame 1, GAC63, HuEL, HUELexpressed in human embryonic lung, Human embryonic lung protein, solute carrier family 30 (zinc transporter), member 9, Solute carrier family 30 member 9, zinc transporter 9, znT-9, ZNT9 | |
SLC30A9 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title