Alfa Aesar™ Zinc trifluoroacetate hydrate

Manufacturer: Alfa Aesar™ 1868614

Quantity 25g

 View more versions of this product

Catalog No. AA1868614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 207801-31-8
    Formula Weight 291.40 (Anhydrous)
    MDL Number MFCD00061595
    UN Number UN3077
    Chemical Name or Material Zinc trifluoroacetate hydrate

    Specifications

    Physical Form Powder/Chunks
    Formula Weight 291.40 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00061595
    Quantity 25g
    UN Number UN3077
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Sensitivity Hygroscopic
    CAS 207801-31-8
    Health Hazard 2 GHS H Statement
    H300
    Fatal if swallowed.
    Chemical Name or Material Zinc trifluoroacetate hydrate

    GHS H Statement
    H300
    Fatal if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit