Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc tungsten oxide, 99.9% (metals basis)

Click to view available options
Quantity:
10g
50g
250g
13597-56-3
O4WZn
313.216
MFCD00049633
LJSXSLGFYGBEPA-UHFFFAOYSA-N
zinc tungsten oxide,zinc tungstate,zinkwolframat,wo4.zn,zinc 2+ tungstate,zinc, ion zn2+ tungstate
22022519
zinc;dioxido(dioxo)tungsten
[O-][W](=O)(=O)[O-].[Zn+2]
13597-56-3
O4WZn
313.216
MFCD00049633
LJSXSLGFYGBEPA-UHFFFAOYSA-N
zinc tungsten oxide,zinc tungstate,zinkwolframat,wo4.zn,zinc 2+ tungstate,zinc, ion zn2+ tungstate
22022519
zinc;dioxido(dioxo)tungsten
[O-][W](=O)(=O)[O-].[Zn+2]
Solubility Information Insoluble in water.
Formula Weight 313.22
Physical Form -200 Mesh Powder
Percent Purity 99.9%
Mesh Size −200 Mesh
Quantity 10g
Chemical Name or Material Zinc tungsten oxide
Assay (metals basis)

GHS H Statement
H335
May cause respiratory irritation.

P261-P271-P304+P340-P312-P501c

H335

EINECSNumber : 237-053-6

TSCA : Yes

Recommended Storage : Ambient temperatures