Learn More
Zincon monosodium salt
Description
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.
Solubility
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∽203°C (decomposition) |
| Quantity | 1 g |
| Beilstein | 3872699 |
| Solubility Information | Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents. |
| Formula Weight | 462.42 |
| Chemical Name or Material | Zincon monosodium salt |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 263-651-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.