Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIP Kinase Rabbit anti-Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206727
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6727 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6727 Supplier Fabgennix Inc Supplier No. DAPK3301AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZIP Kinase Polyclonal antibody specifically detects ZIP Kinase in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ZIP Kinase
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene DAPK3
Gene Accession No. O54784, O88764
Gene Alias DAP kinase 3; DAP3K; Dapk3; Dapkl; DAP-like kinase; death associated protein kinase 3; death-associated kinase 3; Death-associated like kinase; death-associated protein kinase 3; DLK; MYPT1 kinase; ZIP; ZIP kinase; ZIP kinase isoform; Zipk; ZIP-kinase; zipper-interacting protein kinase
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region on 400-448 on mouse Death-associated protein kinase 3 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13144, 64391
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.