Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIP11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120717 Supplier Invitrogen™ Supplier No. PA5120717
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse testis, Rat testis. Immunogen sequence: HLGATEDPQT ALALNLDPAL MKKSDPRDPT SLLFPESELS IRIGSTGLLS DKRENGEVYQ RKKVAATDLA EGVA.

ZIP11, also known as Slc39A11, is a member of the broader ZIP family of multi-transmembrane domain metal ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. Members of the ZIP family generally transport metal ions from the cell exterior or lumen of intracellular organelles into the cytoplasm, as opposed to the ZnT (SLC30) family of zinc transporters that serve to reduce intracellular zinc availability by promoting zinc efflux from cells or into intracellular vesicles. At least three isoforms of ZIP11 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIP11
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.817 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SLC39A11
Gene Accession No. Q6P6S2, Q8BWY7, Q8N1S5
Gene Alias 1810074D23Rik; C17orf26; SLC39A11; solute carrier family 39 (metal ion transporter), member 11; solute carrier family 39 member 11; solute carrier family 39, member 11; zinc transporter ZIP11; ZIP11; ZIP-11; Zrt- and Irt-like protein 11
Gene Symbols SLC39A11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 101-174 of mouse Slc39a11 (NP_001159975.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201266, 287796, 69806
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.