Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113784 Supplier Invitrogen™ Supplier No. PA5113784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MCFRTKLSVS WVPLFLLLSR VFSTETDKPS AQDSRSRGSS GQPADLLQVL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 77%; Horse: 85%; Human: 100%.

The zinc transporter ZIP12, also known as SLC39A12, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP12 is a multipass membrane protein that belongs to the ZIP transporter subfamily LIV-1. Increased levels of ZIP12 have been observed during acute inflammation in the airway and asthma, suggesting that it may be promote increased zinc uptake, as this can reduce inflammation. The zinc content in brains in individuals with schizophrenia is lower than in individuals with other cerebral diseases, and mutations in ZIP12 have been observed in a small group of schizophrenic patients, suggesting that ZIP12 may play a role in this disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIP12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC39A12
Gene Accession No. Q504Y0
Gene Alias AW046938; bA570F3.1; Gm731; LIV-1 subfamily of ZIP zinc transporter 8; LZT-Hs8; Slc39a12; solute carrier family 39 (metal ion transporter), member 12; solute carrier family 39 (zinc transporter), member 12; solute carrier family 39 member 12; Zinc transporter ZIP12; Zip12; ZIP-12; Zrt- and Irt-like protein 12
Gene Symbols SLC39A12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human SLC39A12 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221074
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.