Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIP13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560566 Supplier Invitrogen™ Supplier No. PA560566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TATACRLDNK ESESWGALLS GERLDT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 88%.

The zinc transporter ZIP13, also known as SLC39A13, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP13 is a multipass membrane protein that belongs to the ZIP transporter subfamily LIV-1. Mutations in ZIP13 have recently been shown to cause a spondylocheiro dysplastic form of Ehlers-Danlos syndrome (SCD-EDS), a generalized skeletal dysplasia involving mainly the spine with clinical abnormalities of the hands in addition to EDS-like features. Other experiments have shown that ZIP13 is required for proper connective tissue development and is involved in BMP/TGF- signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIP13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC39A13
Gene Accession No. Q96H72
Gene Alias LIV-1 subfamily of ZIP zinc transporter 9; LZT-Hs9; SLC39A13; solute carrier family 39 (metal ion transporter), member 13; solute carrier family 39 (zinc transporter), member 13; solute carrier family 39 member 13; Zinc transporter ZIP13; ZIP13; ZIP-13; Zrt- and Irt-like protein 13
Gene Symbols SLC39A13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZIP13. Recombinant protein control fragment (Product #RP-99870).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91252
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.